MAT047031203 Matrix Scientific

3-(5,6-Dimethyl-thieno[2,3-d]pyrimidin-4-ylamino)-propionic acid

  • MDL number MFCD03539088
  • Cas Number : 690688-63-2
  • Molecular Weight : 251.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012134.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012134.html
Availability details for
Quantity
  • Availability
  • Add to Cart