MAT047031210 Matrix Scientific

4-(1,3-Dihydro-isoindol-2-yl)-benzoic acid

  • MDL number MFCD00755022
  • Cas Number : 329715-35-7
  • Molecular Weight : 239.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012140.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012140.html
Availability details for
Quantity
  • Availability
  • Add to Cart