MAT047031211 Matrix Scientific

5-(3,4,5-Trimethoxy-phenyl)-[1,3,4]oxadiazole-2-thiol

  • MDL number MFCD01236999
  • Cas Number : 23269-92-3
  • Molecular Weight : 268.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186-188
Country of Origin USA
Product Link http://www.matrixscientific.com/012141.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012141.html
Availability details for
Quantity
  • Availability
  • Add to Cart