MAT047031225 Matrix Scientific

N-(4-Amino-2-methyl-phenyl)-2-pyrrolidin-1-yl-acetamide

  • MDL number MFCD03434388
  • Cas Number : 436090-52-7
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link http://www.matrixscientific.com/012153.html

Safety Information

MSDS http://www.matrixscientific.com/012153.html
Availability details for
Quantity
  • Availability
  • Add to Cart