MAT047031229 Matrix Scientific

(4-Amino-phenyl)-(3-methyl-piperidin-1-yl)-methanone

  • MDL number MFCD04035366
  • Cas Number : 79868-21-6
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012157.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012157.html
Availability details for
Quantity
  • Availability
  • Add to Cart