MAT047031236 Matrix Scientific

4-Amino-N-thiophen-2-ylmethyl-benzamide

  • MDL number MFCD04035387
  • Cas Number : 436095-49-7
  • Molecular Weight : 232.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012163.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012163.html
Availability details for
Quantity
  • Availability
  • Add to Cart