MAT047031267 Matrix Scientific

3-(5-Nitro-1H-benzoimidazol-2-yl)-propionic acid

  • MDL number MFCD03357066
  • Cas Number : 33299-25-1
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-145
Country of Origin USA
Product Link http://www.matrixscientific.com/012191.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012191.html
Availability details for
Quantity
  • Availability
  • Add to Cart