MAT047031280 Matrix Scientific

4-(Thiophene-2-sulfonylamino)-butyric acid

  • MDL number MFCD02333807
  • Cas Number : 379251-18-0
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012203.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012203.html
Availability details for
Quantity
  • Availability
  • Add to Cart