MAT047031290 Matrix Scientific

(3-Ethoxy-propyl)-(phenyl-pyridin-2-yl-methyl)-amine

  • MDL number MFCD05240241
  • Cas Number : 799256-20-5
  • Molecular Weight : 270.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012212.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012212.html
Availability details for
Quantity
  • Availability
  • Add to Cart