MAT047031291 Matrix Scientific

5-(Pyridin-4-ylcarbamoyl)-3H-imidazole-4-carboxylic acid

  • MDL number MFCD04113984
  • Cas Number : 627470-18-2
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012213.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012213.html
Availability details for
Quantity
  • Availability
  • Add to Cart