MAT047031340 Matrix Scientific

4-(3,4-Dimethoxy-phenyl)-5-methyl-2H-pyrazol-3-ylamine

  • MDL number MFCD00439266
  • Cas Number : 347840-95-3
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012257.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012257.html
Availability details for
Quantity
  • Availability
  • Add to Cart