MAT047031357 Matrix Scientific

Furan-2-ylmethyl-phenethyl-amine

  • MDL number MFCD01469634
  • Cas Number : 4439-55-8
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 107�06 Torr
Product Link http://www.matrixscientific.com/012272.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012272.html
Availability details for
Quantity
  • Availability
  • Add to Cart