MAT047031364 Matrix Scientific

alpha-Methyl-2-trifluoromethylbenzyl bromide

  • MDL number MFCD00039226
  • Cas Number : 194152-29-9
  • Molecular Weight : 253.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002212.html

Safety Information

hcodes Corrosive/Lachrymator
MSDS http://www.matrixscientific.com/002212.html
Availability details for
Quantity
  • Availability
  • Add to Cart