MAT047031376 Matrix Scientific

(4-Fluoro-benzyl)-(3-isopropoxy-propyl)-amine

  • MDL number MFCD04113957
  • Cas Number : 799260-49-4
  • Molecular Weight : 225.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012289.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012289.html
Availability details for
Quantity
  • Availability
  • Add to Cart