MAT047031380 Matrix Scientific

3-(Biphenyl-4-sulfonyl)-propionic acid

  • MDL number MFCD03876447
  • Cas Number : 20025-59-6
  • Molecular Weight : 290.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168
Country of Origin USA
Product Link http://www.matrixscientific.com/012293.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012293.html
Availability details for
Quantity
  • Availability
  • Add to Cart