MAT047031397 Matrix Scientific

3-[Thien-2-yl-5-(trifluoromethyl)]isoxazole

  • MDL number MFCD00276611
  • Cas Number : 175203-89-1
  • Molecular Weight : 219.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-59
Country of Origin USA
Product Link http://www.matrixscientific.com/002215.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002215.html
Availability details for
Quantity
  • Availability
  • Add to Cart