MAT047031400 Matrix Scientific

N,N-Dimethyl-N'-pyridin-4-ylmethyl-benzene-1,4-diamine

  • MDL number MFCD04585452
  • Cas Number : 52725-19-6
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012311.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012311.html
Availability details for
Quantity
  • Availability
  • Add to Cart