MAT047031415 Matrix Scientific

3-(4-Methyl-thiazol-2-ylcarbamoyl)-7-oxa-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

  • MDL number MFCD06411476
  • Cas Number : 1005263-26-2
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012325.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012325.html
Availability details for
Quantity
  • Availability
  • Add to Cart