MAT047031427 Matrix Scientific

(Methanesulfonyl-phenethyl-amino)-acetic acid

  • MDL number MFCD04218385
  • Cas Number : 35839-52-2
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012336.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012336.html
Availability details for
Quantity
  • Availability
  • Add to Cart