MAT047031431 Matrix Scientific

4-(4-Methanesulfonyl-piperazin-1-yl)-phenylamine

  • MDL number MFCD03848236
  • Cas Number : 442549-42-0
  • Molecular Weight : 255.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012339.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012339.html
Availability details for
Quantity
  • Availability
  • Add to Cart