MAT047031466 Matrix Scientific

6-Methoxy-2,3,4,9-tetrahydro-carbazol-1-one

  • MDL number MFCD00459564
  • Cas Number : 3449-49-8
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-214
Country of Origin USA
Product Link http://www.matrixscientific.com/012370.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012370.html
Availability details for
Quantity
  • Availability
  • Add to Cart