MAT047031468 Matrix Scientific

3-(4-Oxo-4,5-dihydro-thiazol-2-ylamino)-propionic acid

  • MDL number MFCD04113965
  • Cas Number : 797027-57-7
  • Molecular Weight : 188.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012372.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012372.html
Availability details for
Quantity
  • Availability
  • Add to Cart