MAT047031491 Matrix Scientific

(3,5-Dimethyl-4-nitro-pyrazol-1-yl)-acetic acid

  • MDL number MFCD00297279
  • Cas Number : 345637-69-6
  • Molecular Weight : 199.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012393.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012393.html
Availability details for
Quantity
  • Availability
  • Add to Cart