MAT047031517 Matrix Scientific

Bicyclo[2.2.1]hept-5-en-2-ylmethyl-(tetrahydro-furan-2-ylmethyl)-amine

  • MDL number MFCD06411478
  • Cas Number : 1005263-32-0
  • Molecular Weight : 207.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012417.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012417.html
Availability details for
Quantity
  • Availability
  • Add to Cart