MAT047031526 Matrix Scientific

[5-(4-Amino-phenyl)-furan-2-yl]-methanol

  • MDL number MFCD01952800
  • Cas Number : 54146-51-9
  • Molecular Weight : 189.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012425.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012425.html
Availability details for
Quantity
  • Availability
  • Add to Cart