MAT047031545 Matrix Scientific

(4-Methylsulfanyl-benzyl)-(3-morpholin-4-yl-propyl)-amine

  • MDL number MFCD05238074
  • Cas Number : 726162-72-7
  • Molecular Weight : 280.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012442.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012442.html
Availability details for
Quantity
  • Availability
  • Add to Cart