MAT047031547 Matrix Scientific

(1-Ethyl-pyrrolidin-2-ylmethyl)-thiophen-2-ylmethyl-amine

  • MDL number MFCD05238076
  • Cas Number : 726163-51-5
  • Molecular Weight : 224.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012444.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012444.html
Availability details for
Quantity
  • Availability
  • Add to Cart