MAT047031549 Matrix Scientific

(3,4-Dimethoxy-benzyl)-(1-ethyl-pyrrolidin-2-ylmethyl)-amine

  • MDL number MFCD05238078
  • Cas Number : 726161-90-6
  • Molecular Weight : 278.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012446.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012446.html
Availability details for
Quantity
  • Availability
  • Add to Cart