MAT047031573 Matrix Scientific

4-[4-(2-Fluoro-benzyl)-piperazin-1-yl]-phenylamine

  • MDL number MFCD05237203
  • Cas Number : 709001-17-2
  • Molecular Weight : 285.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link http://www.matrixscientific.com/012467.html

Safety Information

MSDS http://www.matrixscientific.com/012467.html
Availability details for
Quantity
  • Availability
  • Add to Cart