MAT047031582 Matrix Scientific

3-(2-Diethylamino-ethyl)-4-oxo-10-oxa-3-aza-tricyclo[5.2.1.0*1,5*]dec-8-ene-6-carboxylic acid

  • MDL number MFCD06411480
  • Cas Number : 1005060-07-0
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012475.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012475.html
Availability details for
Quantity
  • Availability
  • Add to Cart