MAT047031585 Matrix Scientific

5-[(Diisopropylamino)-methyl]-furan-2-carboxylic acid

  • MDL number MFCD02737449
  • Cas Number : 429651-03-6
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012478.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012478.html
Availability details for
Quantity
  • Availability
  • Add to Cart