MAT047031594 Matrix Scientific

5-Methyl-2-propyl-2H-pyrazol-3-ylamine

  • MDL number MFCD05237221
  • Cas Number : 3524-34-3
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012486.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012486.html
Availability details for
Quantity
  • Availability
  • Add to Cart