MAT048692256 Matrix Scientific

5-Benzyl-2-methoxybenzaldehyde

  • MDL number MFCD06245849
  • Cas Number : 722493-91-6
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55
Country of Origin USA
Boiling point 229-230� Torr
Product Link http://www.matrixscientific.com/015783.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/015783.html
Availability details for
Quantity
  • Availability
  • Add to Cart