MAT048692423 Matrix Scientific

4-Bromotetrafluorophenol

  • MDL number MFCD06246024
  • Cas Number : 1998-61-4
  • Molecular Weight : 244.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 28-30
Purity 97
Storage temp. Ambient
Country of Origin USA
Boiling point 76-77�m
Product Link http://www.matrixscientific.com/012838.html

Safety Information

hcodes Corrosive/Irritant
MSDS http://www.matrixscientific.com/012838.html
Availability details for
Quantity
  • Availability
  • Add to Cart