MAT048693025 Matrix Scientific

3,5-Bis(benzyloxy)benzyl bromide

  • MDL number MFCD02093444
  • Cas Number : 24131-32-6
  • Molecular Weight : 383.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-88
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/012586.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/012586.html
Availability details for
Quantity
  • Availability
  • Add to Cart