MAT048693028 Matrix Scientific

N-(2-Amino-ethyl)-isonicotinamide

  • MDL number MFCD01332312
  • Cas Number : 17704-88-0
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-103
Country of Origin USA
Product Link http://www.matrixscientific.com/012803.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012803.html
Availability details for
Quantity
  • Availability
  • Add to Cart