MAT048693043 Matrix Scientific

4-Chloro-3-formylpyrazole

  • MDL number MFCD00110641
  • Cas Number : 623570-54-7
  • Molecular Weight : 130.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/012591.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012591.html
Availability details for
Quantity
  • Availability
  • Add to Cart