MAT048693059 Matrix Scientific

1-(4-tert-Butylphenyl)-2-(pyrimidin-4-yl)ethanol

  • MDL number MFCD03426054
  • Cas Number : 849021-31-4
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-99
Country of Origin USA
Product Link http://www.matrixscientific.com/007292.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007292.html
Availability details for
Quantity
  • Availability
  • Add to Cart