MAT048694136 Matrix Scientific

3-(Cyclopropylmethoxy)benzaldehyde

  • MDL number MFCD06246720
  • Cas Number : 58986-61-1
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/015678.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/015678.html
Availability details for
Quantity
  • Availability
  • Add to Cart