MAT048694173 Matrix Scientific

2-(Cyclopropylmethoxy)-4-fluorobenzaldehyde

  • MDL number MFCD06246757
  • Cas Number : 883525-53-9
  • Molecular Weight : 194.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/015882.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/015882.html
Availability details for
Quantity
  • Availability
  • Add to Cart