MAT048694709 Matrix Scientific

4-Ethoxy[1,1'-biphenyl]-3-carbaldehyde

  • MDL number MFCD06247318
  • Cas Number : 883535-58-8
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/015744.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/015744.html
Availability details for
Quantity
  • Availability
  • Add to Cart