MAT048694722 Matrix Scientific

4-Ethoxy-2,6-dimethylbenzaldehyde

  • MDL number MFCD06247332
  • Cas Number : 860748-43-2
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-99
Country of Origin USA
Product Link http://www.matrixscientific.com/015716.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/015716.html
Availability details for
Quantity
  • Availability
  • Add to Cart