MAT048694735 Matrix Scientific

2-Ethoxy-4-fluorobenzaldehyde

  • MDL number MFCD06247345
  • Cas Number : 883537-24-4
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/015880.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/015880.html
Availability details for
Quantity
  • Availability
  • Add to Cart