MAT048694972 Matrix Scientific

(1,2-Dimethyl-1H-indol-5-yloxy)-acetic acid

  • MDL number MFCD06011073
  • Cas Number : 25888-11-3
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-199
Country of Origin USA
Product Link http://www.matrixscientific.com/012759.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012759.html
Availability details for
Quantity
  • Availability
  • Add to Cart