MAT048694980 Matrix Scientific

[3-(2,2-Dimethyl-propionyl)-indol-1-yl]-aceticacid

  • MDL number MFCD06010888
  • Cas Number : 842964-33-4
  • Molecular Weight : 259.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012776.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012776.html
Availability details for
Quantity
  • Availability
  • Add to Cart