MAT048694996 Matrix Scientific

2,5-Dichloro-4,6-dimethylnicotinonitrile

  • MDL number MFCD00052631
  • Cas Number : 91591-63-8
  • Molecular Weight : 201.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-87
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/012602.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012602.html
Availability details for
Quantity
  • Availability
  • Add to Cart