MAT048695001 Matrix Scientific

4,5-Dimethoxy-2-methylbenzaldehyde

  • MDL number MFCD00094433
  • Cas Number : 7721-62-2
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76
Storage temp. Ambient/argon
Country of Origin USA
Boiling point 110-111�2 Torr
Product Link http://www.matrixscientific.com/015775.html

Safety Information

hcodes Irritant/Stored under Argon
MSDS http://www.matrixscientific.com/015775.html
Availability details for
Quantity
  • Availability
  • Add to Cart