MAT048696055 Matrix Scientific

5,5,6,6,7,7,7-Heptafluoroheptan-1-ol

  • MDL number MFCD06247732
  • Cas Number : 883543-64-4
  • Molecular Weight : 242.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/012851.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012851.html
Availability details for
Quantity
  • Availability
  • Add to Cart