MAT048696126 Matrix Scientific

8-Iodo-1,1,1,2,2,3,3,4,4-nonafluorooctane

  • MDL number MFCD06247808
  • Cas Number : 99324-98-8
  • Molecular Weight : 402.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 96-97�mm
Product Link http://www.matrixscientific.com/012856.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012856.html
Availability details for
Quantity
  • Availability
  • Add to Cart