MAT048696127 Matrix Scientific

6-Iodo-1,1,1,2,2-pentafluorohexane

  • MDL number MFCD06247809
  • Cas Number : 344452-10-4
  • Molecular Weight : 302.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Boiling point 62-64� Torr
Product Link http://www.matrixscientific.com/012857.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012857.html
Availability details for
Quantity
  • Availability
  • Add to Cart